Order Entry
Puerto Rico
ContactUsLinkComponent
IBI rBAC Mini Total RNA Kit, IBI Scientific
IBI rBAC Mini Total RNA Kit, IBI Scientific
Supplier:  IBI Scientific
Certificates
IBI rBAC Mini Total RNA Kit, IBI Scientific
Supplier:  IBI Scientific

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Bacteria
  • Target molecule:
    RNA
  • Description:
    rBAC Mini Total RNA kit
  • Yield:
    40-50 µg from 1 x 10⁹ Bacterial cells

Specifications