ContactUsLinkComponent
Activate™ High-level Chlorine Test Strips, Deardorff Fitzsimmons Corp
Activate™ High-level Chlorine Test Strips, Deardorff Fitzsimmons Corp
Catalog # 89136-484
CAS Number:  
Activate™ High-level Chlorine Test Strips, Deardorff Fitzsimmons Corp
Catalog # 89136-484
Supplier Number:  77085
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    High-level Chlorine Test Strips
  • Cat. No.:
    89136-484
  • Range:
    1000-2500-5000-7500-10000
  • Supplier no.:
    77085
Specifications
About this item
Activate™ High-level Chlorine Test Strips allow a higher and broader range test for available chlorine, with results in just 30 seconds.
  • Safety checks the strength of diluted bleach in 30 seconds
  • Measures ppm of available chlorine for disinfecting purposes
  • Ranges: 1,000 2,500 5,000 7,500 10,000
  • Desiccant-lined vial helps keep strips fresh
  • Ergonomically designed snap closure
  • 50 strips per vial and 6 vials per case