About this item
Prostate Specific Antigen (PSA) control slides contain sections of positive staining prostate and negative staining lung.
- Tissue type: (+) Prostate / (-) Lung
- All control tissue is sectioned on Superfrost® Plus slides
- Staining validation technical memo included in each set
- All tissues are formalin fixed, paraffin embedded FFPE
- All slides are air dried
- IHC slides are cut at 4 µm
- Standard tissue placement is mid-high on slide
Newcomer supply Prostate Specific Antigen (PSA) control slides are for the positive immunohistochemical staining of PSA, present in prostate tissue and expressed in the vast majority of prostate carcinomas
Staining Results:
PSA positive expression will stain brown cytoplasmic staining.
Lung will stain negative.
Nuclei will stain blue.
These positive control slides are intended to be used to verify histological techniques and reagent reactivity. These slides are to be used for the qualitative purpose of determining positive or negative results, and are not intended to be used for any quantitative purpose.
It recognizes primary and metastatic prostatic neoplasms and rarely in tumors of non-prostatic origin.
10 set = 10 unstained slides and 1 stained slide.
98 set = 98 unstained slides and 2 stained slides. (First & Last Slides are Stained)
Reactivity of IHC control slides are guaranteed for one year from the date of receipt and should be revalidated after one year to verify continued reactivity.
Please note the following: Allow 2-3 weeks for delivery for large sets. In human tissue unless otherwise indicated.