Accessories for BioSpectrometer®, Basic and Kinetic, Eppendorf
75875-044
- Description:Eppendorf BioSpectrometer® Reference Filter Set
- Cat. No.:75875-044
- Supplier no.:6135928001
- Details:For verifying photometric accuracy and wavelength systematic error (according to NIST®)