ContactUsLinkComponent
Accessories for BioPhotometer® D30, Eppendorf
Accessories for BioPhotometer® D30, Eppendorf
  75875-040
 :  Eppendorf
undefined
Accessories for BioPhotometer® D30, Eppendorf
  75875-040
 :  Eppendorf
 :  6133928004
  • Description:
    Eppendorf BioPhotometer® D30 Reference Filter Set, filter set for verification of wavelength accuracy and precision at 260 nm and 280 nm
  • Cat. No.:
    75875-040
  • Supplier no.:
    6133928004