ContactUsLinkComponent
Accessories for BioSpectrometer® Fluorescence, Eppendorf
Accessories for BioSpectrometer® Fluorescence, Eppendorf
Catalog # 75875-046
Supplier:  Eppendorf
undefined
Accessories for BioSpectrometer® Fluorescence, Eppendorf
Catalog # 75875-046
Supplier:  Eppendorf
Supplier Number:  6137928009
Specifications
  • Description:
    Eppendorf BioSpectrometer® fluorescence Reference Filter Set, for verifying photometric accuracy and wavelength systematic error and verifying fluorimetric precision and linearity
  • Cat. No.:
    75875-046
  • Supplier no.:
    6137928009