MagID Magnetic Isolation Device, OZ Biosciences
Catalog # 103258-106
Supplier: OZ Biosciences
CAS Number:
Specifications
- Description:OZ Biosciences MagID, Magnetic Isolation Device, Holds 1.5 mL Tubes
- Cat. No.:103258-106
- Supplier no.:DM30000
Specifications
About this item
OZ Biosciences MagID Magnetic Isolation Device is ideal for coupling reactions, washing procedures, aspiration, pipetting etc.
- Ideal for your magnetic nanoparticles coupling reaction and purification
- Adapted to working solutions ranging from 10 µL to 2 mL
- Perfect for a quick magnetic separation process (< 5 minutes) with a high yield separation
- Durable and easy to use with an open faced design