Order Entry
Puerto Rico
ContactUsLinkComponent
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) solution 10%, PROTEIN GRADE® detergent, sterile-filtered, Calbiochem®, Millipore®
Corrosive
Environment
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) solution 10%, PROTEIN GRADE® detergent, sterile-filtered, Calbiochem®, Millipore®
Catalog # 80057-666
Supplier:  MilliporeSigma
CAS Number:  
Corrosive
Environment
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) solution 10%, PROTEIN GRADE® detergent, sterile-filtered, Calbiochem®, Millipore®
Catalog # 80057-666
Supplier:  MilliporeSigma
Supplier Number:  648463-50ML
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic ampoule
  • Size:
    50 ml
  • Cat. No.:
    80057-666
  • Supplier no.:
    648463-50ML

Specifications

About this item

 :
Storage Temperature:Refrigerator
UN:3082
ADR:9,III
 :GHS05
 :GHS09
CAS Number:9002-93-1
 :9036-19-5
MDL Number:MFCD00132505