ContactUsLinkComponent
Laboratory Dry Sterilizer with Overheat Protection, Yamato
undefined
Laboratory Dry Sterilizer with Overheat Protection, Yamato
  • Catalog No:
  • 75846-698
  • 75846-700
  • Control Type:
  • Digital PID Controller
  • Digital PID Controller
  • Max. Load Per Shelf:
  • ~15 kg
  • ~15 kg
  • Programs:
  • Maximum 99 Steps, 99 Patterns, Repeat Operation
  • Maximum 99 Steps, 99 Patterns, Repeat Operation
  • Sensor:
  • K Type Thermocouple
  • K Type Thermocouple
  • Timer:
  • 0 Minutes to 99 Hours, 59 Minutes (Increment: 1 Minute or 1 Hour )
  • 0 Minutes to 99 Hours, 59 Minutes (Increment: 1 Minute or 1 Hour )