Order Entry
Puerto Rico
ContactUsLinkComponent
Barium, Ion chromatography standard solution, Specpure®, Ba2+ 1000 ug/ml, Thermo Scientific
Barium, Ion chromatography standard solution, Specpure®, Ba2+ 1000 ug/ml, Thermo Scientific
Catalog # AA47208-AE
CAS Number:  
Barium, Ion chromatography standard solution, Specpure®, Ba2+ 1000 ug/ml, Thermo Scientific
Catalog # AA47208-AE
Supplier Number:  047208-AE
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    H₂O
  • Pack type:
    Glass Bottle
  • Size:
    100 ml
  • Cat. No.:
    AA47208-AE
  • Element:
    Barium
  • Supplier no.:
    47208-AE

Specifications

About this item

Barium nitrate in water