Lithium, Ion Chromatography Standard Solution, Specpure®, Li+ 1000 ug/ml, Thermo Scientific
Catalog # AA47252-AE
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Concentration:1000 µg/ml
- Matrix:H₂O
- Pack type:Glass Bottle
- Size:100 ml
- Cat. No.:AA47252-AE
- Element:Lithium
- Supplier no.:47252-AE