Order Entry
Puerto Rico
ContactUsLinkComponent
2-Mercapto-1-methylimidazole USP Reference Standard
Health
Irritant
2-Mercapto-1-methylimidazole USP Reference Standard
Catalog # 101095-470
Supplier:  USP
CAS Number:  
Health
Irritant
2-Mercapto-1-methylimidazole USP Reference Standard
Catalog # 101095-470
Supplier:  USP
Supplier Number:  1411005
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    200 mg
  • Cat. No.:
    101095-470
  • Supplier no.:
    1411005

Specifications

About this item

 :
 :114.17 g/mol
 :GHS08
 :GHS07
CAS Number:60-56-0
Formula:C₄H₆N₂S
MDL Number:MFCD00179321