ContactUsLinkComponent
EnzyChrom™ Pyruvate Assay Kit, BioAssay Systems
EnzyChrom™ Pyruvate Assay Kit, BioAssay Systems
Catalog # 75878-166
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
EnzyChrom™ Pyruvate Assay Kit, BioAssay Systems
Catalog # 75878-166
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  EPYR-100
CAS Number:  
Specifications
  • Description:
    Pyruvate Assay Kit
  • Size:
    100 assays
  • Method:
    OD570 nm, or FL530/585 nm
  • Sample:
    Biological
  • Processing Time:
    20 minutes
  • Cat. No.:
    75878-166
  • Supplier no.:
    EPYR-100
Specifications
About this item
For quantitative determination of pyruvate and evaluation of drug effects on its metabolism.
  • Linear detection range 2 - 500 μM pyruvate for colorimetric assays and 0.2 - 50 μM for fluorimetric assays
  • Convenient room temperature, high-throughput assay
Pyruvate is a key intermediate in cellular metabolic pathways
BioAssay Systems’ pyruvate assay uses a single Working Reagent that combines pyruvate oxidase and hydrogen peroxide determination in one step. The color intensity of the reaction product at 570nm or fluorescence intensity at λex/em = 530/585nm is directly proportional to pyruvate concentration in the sample.
Pyruvate can be converted to carbohydrates via gluconeogenesis, to fatty acids or energy through acetyl-CoA, to the amino acid alanine and to ethanol. Abnormal levels of pyruvate have been linked to liver diseases and metabolic disorders. Simple, direct and automation-ready. Procedures for measuring pyruvate concentrations find wide applications in research and drug discovery.