Specifications
- Description:Ketone Body Assay Kit
- Size:100 assays
- Method:OD340 nm
- Sample:Serum, plasma, urine etc.
- Processing Time:30 minutes
- Cat. No.:75878-144
- Supplier no.:EKBD-100
Specifications
About this item
For quantitative determination of ketone body (acetoacetate and 3-hydroxybutyrate) and evaluation of drug effects on its metabolism.
- Linear detection range of 0.12 to 8 mM for each ketone body in 96-well plate assay
- The procedure involves adding a single working reagent, and reading the optical density at room temperature
- Can be automated to test many samples per day
Ketone bodies (acetoacetic acid and 3-hydroxybutyric acid) are produced in the liver mainly from oxidation of fatty acids, and are normally present at low concentrations in urine and blood
BioAssay Systems’ EnzyChrom™ ketone body assay is based on 3-hydroxybutyrate dehydrogenase catalyzed reactions, in which the change in NADH absorbance, measured at 340nm, is directly related to the AcAc and BOH concentrations,
Increased ketone concentrations in the blood may lead to metabolic acidosis, which has been associated with diabetes, childhood hypo-glycaemia, growth hormone deficiency, alcohol or salicylate intoxication and inborn errors of metabolism. Simple, direct and automation-ready. Procedures for measuring acetoacetic acid (AcAc) and 3-hydroxybutyric acid (BOH) are very desirable.