Specifications
- Description:HDL/LDL Assay Kit
- Size:100 assays
- Method:OD340 nm
- Species Reactivity:All
- Sample:Serum
- Processing Time:60 minutes
- Cat. No.:75878-140
- Supplier no.:EHDL-100
Specifications
About this item
For quantitative colorimetric determination of HDL and LDL/VLDL and evaluation of drug effects on HDL and LDL/VLDL metabolism.
- Linear detection range 5 - 300 mg/dL cholesterol in 96-well plate assay
- Convenient high-throughput assay
- No 37°C heater is needed
Cholesterol concentrations in High-Density Lipoprotein (HDL) and Low-Density (LDL)/Very-Low-Density (VLDL) Lipoproteins are strong predictors for coronary heart disease
BioAssay Systems’ HDL and LDL/VLDL quantification kit is based on our improved PEG precipitation method in which HDL and LDL/VLDL are separated, and cholesterol concentrations are determined using cholesterol esterase/cholesterol dehydrogenase reagent. In this reaction, NAD is reduced to NADH. The optical density of the formed NADH at 340 nm is directly proportionate to the cholesterol concentration in the sample.
Functional HDL offers protection by removing cholesterol from cells and atheroma. Higher concentrations of LDL and lower concentrations of functional HDL are strongly associated with cardiovascular disease due to higher risk of atherosclerosis. The balances between high- and low-density lipoproteins are solely genetically determined, but can be changed by medications, food choices and other factors. Simple, direct and automation-ready. Procedures for measuring HDL and LDL/VLDL concentrations are very desirable.