Specifications
- Description:Glutamate Assay Kit
- Size:100 assays
- Method:OD565 nm
- Sample:Serum, plasma, urine, cell, tissue etc.
- Processing Time:30 minutes
- Cat. No.:75878-130
- Supplier no.:EGLT-100
Specifications
About this item
For quantitative determination of glutamate and evaluation of drug effects on glutamate metabolism.
- Linear detection range 50 μM to 2.5 mM glutamate
- Convenient room temperature assay. No 37°C heater is needed
- Can be readily automated to test thousands of samples per day
Glutamate is an important chemical in general metabolism
BioAssay Systems’ EnzyChrom™ glutamate assay kit is based on glutamate dehydrogenase catalyzed oxidation of glutamate, in which the formed NADH reduces a formazan (MTT) Reagent. The intensity of the product color, measured at 565 nm, is proportionate to the glutamate concentration in the sample.
It is also a crucial mammalian neurotransmitter that is believed to be involved in a number of neurological and psychiatric disorders such as lateral sclerosis, lathyrism, autism and Alzheimer’s disease. Glutamate is also widely used as a flavor enhancer in the food industry. Simple, direct and automation-ready. Procedures for measuring glutamate concentration are very desirable.