Order Entry
Puerto Rico
ContactUsLinkComponent
 
EnzyFluo™ ERK Phosphorylation Assay Kit, BioAssay Systems
Catalog # 75878-108
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
EnzyFluo™ ERK Phosphorylation Assay Kit, BioAssay Systems
Catalog # 75878-108
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  EERK-100
CAS Number:  

Specifications

  • Description:
    ERK Phosphorylation Assay Kit
  • Size:
    100 assays
  • Method:
    FL360/450 nm, 530/585 nm
  • Sample:
    Cells
  • Processing Time:
    Eight hours
  • Cat. No.:
    75878-108
  • Supplier no.:
    EERK-100
  • Species reactivity:
    Human, mouse, rat

Specifications

About this item

For quantitative fluorescent immunoenzymatic assay of ERK1/2 phosphorylation status in cultured cells.

  • Safe
  • Non-radioactive assay
  • Total and pERK can be measured in the same sample

The mitogen-activated protein kinase (MAPK/ERK) pathway plays a key role in cell proliferation, differentiation and migration

BioAssay Systems’ cell-based ELISA measures dually phosphorylated ERK1/2 in whole cells and normalizes the signal to the total protein content. This simple and efficient assay eliminates the need for cell lysate preparation and can be used to study kinase signaling and the effects of kinase inhibitors on cells. In this assay, cells are grown in 96-well plates and treated with ligands or drugs. Cells are then fixed and permeabilized in the wells. ERK1/2 phosphorylation (pERK) using a fluorescent ELISA followed by total protein measurement in each well.

Stimulation by mitogens eventually leads to phosphorylation of ERK1 (T202/Y204) and ERK2 (T185/Y187). The MAPK/ERK cascade presents many interesting drug targets for the development of cancer therapies.