ContactUsLinkComponent
EnzyChrom™ Lactate Assay Kit, BioAssay Systems
  75878-082
 :  
undefined
EnzyChrom™ Lactate Assay Kit, BioAssay Systems
  75878-082
 :  EDLC-100
 :  
  • Description:
    Lactate Assay Kit
  • Size:
    100 assays
  • Method:
    OD565 nm
  • Sample:
    Serum, plasma, cell culture media etc.
  • Processing Time:
    20 minutes
  • Cat. No.:
    75878-082
  • Supplier no.:
    EDLC-100
For quantitative colorimetric determination of D-lactate (D-lactic acid) and evaluation of drug effects on its metabolism.
  • Linear detection range 0.05 - 2 mM D-lactate in 96-well plate assay
  • Convenient room temperature assay. No 37°C heater is needed
  • Can be readily automated to process thousands of samples per day
Lactate is generated by lactate dehydrogenase (LDH) under hypoxic or anaerobic conditions. Monitoring lactate levels is, therefore, a good indicator of the balance between tissue oxygen demand and utilization and is useful when studying cellular and animal physiology. D-lactate is produced in only minor quantities in animals and measuring for D-lactate in animal samples is a means to determine the presence of bacterial infection. Simple, direct and automation-ready. Procedures for measuring lactate concentration are very desirable. BioAssay Systems’ EnzyChrom™ lactate assay kit is based on lactate dehydrogenase catalyzed oxidation of lactate, in which the formed NADH reduces a formazan (MTT) Reagent. The intensity of the product color, measured at 565 nm, is proportionate to the lactate concentration in the sample.