Order Entry
Puerto Rico
ContactUsLinkComponent
 
Centramate™ and Centramate™ PE, Lab Tangential Flow Systems, Cytiva (Formerly Pall Lab)
 :  Cytiva
 
undefined
Centramate™ and Centramate™ PE, Lab Tangential Flow Systems, Cytiva (Formerly Pall Lab)
 :  Cytiva

 

  • Catalog No:
  • 29301-353
  • 29301-336
  • 29301-332
  • Type:
  • Centramate PE
  • Centramate
  • Centramate
  • Operating Temperature:
  • 60 °C (140 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • Dimensions:
  • 9.4 x 25.1 x 19.3 cm (3.7 x 9.4 x 7.6 in.)
  • 13.2 x 22.6 x 16.7 cm (5.2 x 8.9 x 6.6 in.)
  • 13.2 x 22.6 x 16.7 cm (5.2 x 8.9 x 6.6 in.)
  • Weight:
  • 6.4 kg (14 lb.)
  • 7.7 kg (17 lb.)
  • 7.7 kg (17 lb.)
  • Hold-up Volume:
  • < 20 mL (per cassette)
  • < 20 mL (per cassette)
  • < 20 mL (per cassette)
  • Operating Pressure:
  • 0.3 bar (30 kPa, 5 psi) – 5.2 bar (520 kPa, 75 psi)
  • 0.3 bar (30 kPa, 5 psi) – 5.2 bar (520 kPa, 75 psi)
  • 0.3 bar (30 kPa, 5 psi) – 5.2 bar (520 kPa, 75 psi)