ContactUsLinkComponent
EnzyChrom™ Ascorbic Acid Assay Kit, BioAssay Systems
EnzyChrom™ Ascorbic Acid Assay Kit, BioAssay Systems
  75878-044
 :  
undefined
EnzyChrom™ Ascorbic Acid Assay Kit, BioAssay Systems
  75878-044
 :  EASC-100
 :  
  • Description:
    Ascorbic Acid Assay Kit
  • Size:
    100 assays
  • Method:
    OD570 nm, or FL530/585 nm
  • Sample:
    Serum, plasma, urine, saliva, milk, tissue, and cell culture.
  • Processing Time:
    10 minutes
  • Cat. No.:
    75878-044
  • Supplier no.:
    EASC-100
For quantitative determination of ascorbate (ascorbic acid) and evaluation of drug effects on ascorbic acid metabolism.
  • Linear detection range colorimetric assay 6 to 1000 μM, fluorimetric assay 1 to 100 μM ascorbic acid
Ascorbic acid (the L-enantiomer commonly known as vitamin C) is an important antioxidant found in living organisms and applied as additives in food and other industrial processes
BioAssay Systems’ method provides a simple, direct and high-throughput assay for measuring ascorbic acid. In this assay, ascorbic acid is oxidized by ascorbate oxidase resulting in the production of H2O2 which reacts with a specific dye to form a pink colored product. The color intensity at 570nm or fluorescence intensity (530/585 nm) is directly proportional to the ascorbic acid concentration in the sample.
By reacting with reactive oxygen species, it protects the cell from oxidative damages.