To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Sialic acid Assay Kit
- Size:100 assays
- Environmentally Preferable:
- Method:OD549nm, or FL555/585 nm
- Sample:Serum, plasma, saliva, milk, etc.
- Processing Time:2.5 hours
- Brand Name:QuantiChrom™
- Cat. No.:75878-330
- Supplier no.:DSLA-100
Specifications
About this item
For quantitative determination of free and total sialic acid (NANA) and evaluation of drug effects on sialic acid
- Sensitive and accurate. Use as little as 60 μL samples.
- Linear detection range in 96-well plate: 5 to 1000 μM sialic acid for colorimetric assays and 0.5 to 100 μM for fluorimetric assays.
- Method: OD549nm, or FL555/585nm
- Samples: Serum, plasma, saliva, milk etc
SIALIC ACID is a general name for nine carbon acidic sugars with N- or O-substituted derivatives. The most common member of these sugars is N-acetylneuraminic acid (NANA). Sialic acid is widely distributed throughout mammalian tissues and fluids including serum. Sialylated oligosaccharides have been shown to exhibit antiviral properties and are also known to influence blood coagulation and cholesterol levels. The sialic acid level in body fluids is also an important marker for diagnosing cancer. Simple and direct procedures for measuring sialic acid concentrations find wide applications in research and drug discovery. BioAssay Systems’ sialic acid assay uses an improved Warren method, in which sialic acid is oxidized to formylpyruvic acid which reacts with thiobarbituric acid to form a pink colored product. The color intensity at 549 nm or fluorescence intensity at λex/em = 555/585 nm is directly proportional to sialic acid concentration in the sample.