HiVac Valves with Straight PTFE Tip, Chemglass
Catalog # 89094-262
Supplier: Chemglass
Small Business Enterprise
Specifications
- Cat. No.:89094-262
- Bore size:0 - 4 mm PTFE stopcock
- Supplier no.:CGK-553-01
Specifications
About this item
These high vacuum valves feature a barrel with two sidearms attached at a 180° angle to each other, on the same plane.