Order Entry
Puerto Rico
ContactUsLinkComponent
QuantiChrom™ Sorbitol Dehydrogenase Assay Kit, BioAssay Systems
QuantiChrom™ Sorbitol Dehydrogenase Assay Kit, BioAssay Systems
Catalog # 75878-326
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
QuantiChrom™ Sorbitol Dehydrogenase Assay Kit, BioAssay Systems
Catalog # 75878-326
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  DSDH-100
CAS Number:  

Specifications

  • Description:
    Sorbitol Dehydrogenase Assay Kit
  • Size:
    100 assays
  • Method:
    OD565 nm
  • Sample:
    Biological Samples e.g. plasma, serum, urine, tissue and culture media.
  • Processing Time:
    15 minutes
  • Cat. No.:
    75878-326
  • Supplier no.:
    DSDH-100

Specifications

About this item

For quantitative determination of sorbitol dehydrogenase enzyme activity.

  • Linear detection range 0.1 - 125 U/L sorbitol dehydrogenase
  • Can be readily automated to process thousands of samples per day

Sorbitol dehydrogenase (SDH) is an enzyme that catalyzes the interconversion of sorbitol and fructose. Elevated blood serum SDH levels indicate liver damage; thus, SDH plays an important role in the diagnosis of liver disease, especially in combination with aminotransferases. SDH levels are also measured to evaluate diabetic complications such as proliferative diabetic retinopathy. BioAssay Systems’ non-radioactive, colorimetric SDH assay is based on the reduction of the tetrazolium salt MTT in a NADH-coupled enzymatic reaction to a reduced form of MTT which exhibits an absorption maximum at 565 nm. The increase in absorbance at 565 nm is directly proportional to the enzyme activity.