To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Description:Albumin (BCP) Assay Kit
- Size:250 assays
- Method:OD610 nm (BCP)
- Sample:Serum, plasma, urine, biological preparations
- Processing Time:Five minutes
- Cat. No.:75877-982
- Supplier no.:DIAP-250
For quantitative determination of albumin concentrations (BCP method).
- Detection range 0.3 g/dL (45 μM) to 5 g/dL (750 μM) albumin in 96-well plate assay
- The procedure involves addition of a single working reagent and incubation for five minutes
- Can readily automated for processing thousands of samples per day
Albumin is the most abundant plasma protein in human
BioAssay Systems’ BCP albumin assay kit is designed to measure albumin directly in biological samples without any pretreatment. The improved method utilizes bromcresol purple that forms a colored complex specifically with albumin. The intensity of the color, measured at 610nm, is directly proportional to the albumin concentration in the sample. The optimized formulation substantially reduces interference by substances in the raw samples.
It accounts for about 60% of the total serum protein. Albumin plays important physiological roles, including maintenance of colloid osmotic pressure, binding of key substances such as long-chain fatty acids, bile acids, bilirubin, haematin, calcium, magnesium. It has anti-oxidant and anticoagulant effects, and also acts as a carrier for nutritional factors and drugs, as an effective plasma pH buffer. Serum albumin is a reliable prognostic indicator for morbidity and mortality, liver disease, nephritic syndrome, malnutrition and protein-losing enteropathies. High levels are associated with dehydration. Simple, direct and automation-ready. Procedures for measuring albumin concentration in biological samples are becoming popular in Research and Drug Discovery.