Order Entry
Puerto Rico
ContactUsLinkComponent
QuantiChrom™ Glucose-6-Phosphate Dehydrogenase Kit, BioAssay Systems
QuantiChrom™ Glucose-6-Phosphate Dehydrogenase Kit, BioAssay Systems
Catalog # 75877-978
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
QuantiChrom™ Glucose-6-Phosphate Dehydrogenase Kit, BioAssay Systems
Catalog # 75877-978
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  DGPDH-100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Glucose-6-Phosphate Dehydrogenase Kit
  • Size:
    100 assays
  • Method:
    OD460 nm
  • Sample:
    Plasma, serum, tissue and culture media, etc.
  • Processing Time:
    20 minutes
  • Cat. No.:
    75877-978
  • Supplier no.:
    DGPDH-100

Specifications

About this item

For quantitative determination of glucose-6-phosphate dehydrogenase activity.

  • Linear detection range 0.2 to 100 U/L
  • Can be readily automated for processing thousands of samples per day

Glucose-6-phosphate dehydrogenase (G6PDH) is a cytosolic enzyme in the pentose phosphate pathway which supplies reducing energy to cells by maintaining the level of the co-enzyme nicotinamide adenine dinucleotide phosphate (NADPH). G6PDH reduces nicotinamide adenine dinucleotide phosphate (NADP) to NADPH while oxidizing glucose-6-phosphate (G6P). Humans with a genetic deficiency of G6PDH are predisposed to non-immune hemolytic anemia. BioAssay Systems’ non-radioactive, colorimetric G6PDH assay is based on the reduction of the tetrazolium salt MTT in a NADPH-coupled enzymatic reaction to a reduced form of MTT which exhibits an absorption maximum at 565 nm. The increase in absorbance at 565 nm is proportional to the enzyme activity.