About this item
These 17 mm syringe filters are ideal for coarse/crude samples or higher detection limit applications such as food, beverage, or environmental analysis.
- Ideal for routine QA/QC analysis
Nylon Syringe Filters: Ideal for aqueous (non-acidic) or organic sample preparation and HPLC, GC, or dissolution sample analysis.
PTFE (Hydrophobic) Syringe Filters: Versatile filters for use with aggressive organic solvent-based solutions, and especially ideal for HPLC sample preparation. Hydrophobic PTFE syringe filters have broad chemical compatibility and high pH resistance.
PVDF (Hydrophobic) Syringe Filters: Ideal for a wide range of mild organic solutions.
PTFE (Hydrophylic) Syringe Filters: Versatile filters for use with aqueous and aggressive organic solvent-based solutions, and especially ideal for HPLC sample preparation. They have broad chemical compatibility and high pH resistance.