ContactUsLinkComponent
PTFE Tape, Chemglass
PTFE Tape, Chemglass
  16002-900
 :  Chemglass
Small Business Enterprise
undefined
PTFE Tape, Chemglass
  16002-900
 :  Chemglass
 :  CG-1963-B-39
Small Business Enterprise
  • Width:
    1.9 cm (¾")
  • Description:
    PTFE Tape
  • Length:
    13.2 m (43.3')
  • Environmentally Preferable:
  • Cat. No.:
    16002-900
  • Supplier no.:
    CG-1963-B-39
PTFE tape may be used to create a leak-tight seal.