Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Razors and Blades
Blades for Easycut™ Self Retracting Cutter, Cosco
Blades for Easycut™ Self Retracting Cutter, Cosco
Blades for Easycut™ Self Retracting Cutter, Cosco
500000-744
:
Janitorial Supplies
Blades for Easycut™ Self Retracting Cutter, Cosco
500000-744
:
Janitorial Supplies
:
COS091509
Color:
Blue/Black
Description:
Replacement blades
Cat. No.:
500000-744
For use with:
Cosco Easycut™ Self Retracting Cutter (COS-091508, COS-091524)
Supplier no.:
COS091509