Mayo-Stille Dissecting Scissors, OR Grade, Sklar
Supplier: Sklar
Certificates
About this item
Mayo-Stille Dissecting Scissors have rounded edges to permit deep dissection of tough tissue.
Censitrac Ready products are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.