
Silver nitrate 0.1 M AgNO3 (0.1N) volumetric solution, Fluka™
Catalog # BJ72271-1L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:1 L
- Cat. No.:BJ72271-1L
- Supplier no.:72271-1L
Specifications
Specification Test Results
- Assay:0.0998 - 0.1002 mol/l
- Calibrated Against:sodium chloride
Specification Test Results
About this item
:
:169.87 g/mol
Boiling Pt:100 °C (1.013 hPa)
Boiling Pt:100 °C
Melting Pt:<0 °C
Density:1.010 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3264
ADR:8,III
:GHS05
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414
ADR:8,II