Order Entry
Puerto Rico
ContactUsLinkComponent
Sodium hydroxide 1 M NaOH (1N) ≥99.5% volumetric solution, Fluka™
Corrosive
Sodium hydroxide 1 M NaOH (1N) ≥99.5% volumetric solution, Fluka™
Catalog # BJ71463-1L
CAS Number:  
Corrosive
Sodium hydroxide 1 M NaOH (1N) ≥99.5% volumetric solution, Fluka™
Catalog # BJ71463-1L
Supplier Number:  71463-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 L
  • Cat. No.:
    BJ71463-1L
  • Supplier no.:
    71463-1L

Specifications

Specification Test Results

  • Calibrated Against:
    potassium hydrogen phthalate
  • Assay:
    0.998 - 1.002 mol/l

Specification Test Results

About this item

 :
 :40 g/mol
Boiling Pt:100 °C
Melting Pt:0 °C
Density:1.04 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548