
Sulfuric acid concentrate for dilution to 1L, 0.25 M H2SO4 (0.5N) volumetric solution, Fluka™
Catalog # BJ38295-1EA
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Ampoule
- Size:1 Each
- Cat. No.:BJ38295-1EA
- Supplier no.:38295-1EA
Specifications
Specification Test Results
- Factor:0.998 - 1.002
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
About this item
:
:98.08
Boiling Pt:>100 °C (1.013 hPa)
Density:1.180 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2796
ADR:8,II
:GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589