Order Entry
Puerto Rico
ContactUsLinkComponent
Sulfuric acid concentrate for dilution to 1 L, 1 M H2SO4 (2.0 N) volumetric solution, Fluka™
Corrosive
Sulfuric acid concentrate for dilution to 1 L, 1 M H2SO4 (2.0 N) volumetric solution, Fluka™
Catalog # BJ38291-1EA
CAS Number:  
Corrosive
Sulfuric acid concentrate for dilution to 1 L, 1 M H2SO4 (2.0 N) volumetric solution, Fluka™
Catalog # BJ38291-1EA
Supplier Number:  38291-1EA
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 Each
  • Cat. No.:
    BJ38291-1EA
  • Supplier no.:
    38291-1EA

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

 :
 :98.08
Boiling Pt:>100 °C (1.013 hPa)
Density:1.180 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2796
ADR:8,II
 :GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589