

Hydrochloric acid concentrate for dilution to 2L, 1 M HCl (1N) standard volumetric solution, Fluka™
Catalog # BJ38281-1EA
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:1 Each
- Cat. No.:BJ38281-1EA
- Supplier no.:38281-1EA
Specifications
Specification Test Results
- Factor:0.998 - 1.002
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
About this item
:
Boiling Pt:100 °C
Melting Pt:0 °C
Density:1.11 g/cm³
Storage Temperature:Ambient
ADR:8,II
:36.46
UN:1789
:GHS05
:GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324