
Hydrochloric acid concentrate for dilution to 1 L, 0.1 M HCl (0.1 N) volumetric solution, Fluka™
BJ38280-1EA
:
- Pack type:Ampoule
- Size:1 Each
- Cat. No.:BJ38280-1EA
- Supplier no.:38280-1EA
- Factor:0.998 - 1.002
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
:
:100 °C (1.013 hPa)
:0 °C
:1.030 g/cm³ (20 °C)
:Ambient
:8,III
:36.46
:1789
:GHS05
:7647-01-0
:HCl
:MFCD00011324