
Sodium hydroxide concentrate for dilution to 10L, 1 M NaOH (1N) standard volumetric solution, Fluka™
Catalog # BJ38214-1EA
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:1 Each
- Cat. No.:BJ38214-1EA
- Supplier no.:38214-1EA
Specifications
Specification Test Results
- Factor:0.998 - 1.002
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
About this item
:
:40 g/mol
Boiling Pt:120 °C
Melting Pt:319 °C
Density:1.350 g/cm³
Storage Temperature:Ambient
UN:1824
ADR:8,II
:GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548