Order Entry
Puerto Rico
ContactUsLinkComponent
Iodine concentrate for dilution to 0.25 M (0.5 N), Fluka™
Irritant
Health
Iodine concentrate for dilution to 0.25 M (0.5 N), Fluka™
Catalog # BJ38084-1EA
CAS Number:  
Irritant
Health
Iodine concentrate for dilution to 0.25 M (0.5 N), Fluka™
Catalog # BJ38084-1EA
Supplier Number:  38084-1EA
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 Each
  • Cat. No.:
    BJ38084-1EA
  • Supplier no.:
    38084-1EA

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002

Specification Test Results

About this item

 :
 :253.81 g/mol
Melting Pt:113.5 °C
Density:4.930 g/cm3
Storage Temperature:Ambient
UN:3495
ADR:8,III
 :GHS07
 :GHS08
CAS Number:7553-56-2
Formula:I₂
MDL Number:MFCD00011355