
Acetic acid concentrate for dilution to 1L, 0.1 M CH3COOH (0.1N) volumetric solution, Fluka™
BJ38050-1EA
:
- Pack type:Ampoule
- Size:1 Each
- Cat. No.:BJ38050-1EA
- Supplier no.:38050-1EA
- Factor:0.998 - 1.002
:
:60.05 g/mol
:Ambient
:2790
:8,III
:GHS05
:64-19-7
:H₃CCOOH
:MFCD00036152