Ammonium thiocyanate concentrate for dilution to 1L, 0.1 M NH4SCN (0.1N) volumetric solution, Fluka™
Catalog # BJ38020-1EA
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Ampoule
- Size:1 Each
- Cat. No.:BJ38020-1EA
- Supplier no.:38020-1EA
Specifications
Specification Test Results
- Factor:0.998 - 1.002
Specification Test Results
About this item
:
:76.12 g/mol
Boiling Pt:100 °C (1.013 hPa)
Storage Temperature:Ambient
CAS Number:1762-95-4
Formula:NH₄SCN
MDL Number:MFCD00011428