Hydrochloric acid 0.01 M HCl (0.01N) volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Boiling Pt:100 °C
Melting Pt:0 °C
Density:1.0 g/cm3
Storage Temperature:Ambient
EINECS:231-791-2
UN:1789
ADR:8,III
:36.46 g/mol
Specification Test Results
- Factor:0.95 - 1.05
- Ammonium (NH₄):Max. 3 ppm
- Iron (Fe):Max. 0.1 ppm
- Heavy metals (as Pb):Max. 0.5 ppm
- Sulfate (SO₄):Max. 1 ppm