
Hydrochloric acid 2.0 M HCl (2.0N) volumetric solution, Fluka™
:7647-01-0
:
:HCl
:MFCD00011324
:100 °C
:0 °C
:1.04 g/cm³
:Ambient
:1789
:8,III
:36.46
:GHS05
- Factor:0.999 - 1.001
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective