
Hydrochloric acid 0.05 M (0.05 N) volumetric solution, Fluka™
Catalog # BJ35320-1L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:1 L
- Cat. No.:BJ35320-1L
- Supplier no.:35320-1L
Specifications
Specification Test Results
- Vapor Pressure:190 mmHg ( 25 °C)
- Factor:0.998 - 1.002
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
About this item
:
Boiling Pt:100 °C
Melting Pt:0 °C
Density:1.0 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8,III
:36.46
EINECS:231-791-2
UN:1789
:GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324