
Sodium hydroxide 0.1 M NaOH (0.1N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:1,390 °C
Melting Pt:319 °C
Density:2.13 g/cm3
Storage Temperature:Ambient
EINECS:231-791-2
UN:1824
ADR:8,II
:GHS05
Specification Test Results
- Factor:0.999 - 1.001
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective