
Sodium hydroxide 2 M NaOH (2N) volumetric solution, Fluka™
Catalog # BJ-35254-1L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:1 L
- Cat. No.:BJ-35254-1L
Specifications
Specification Test Results
- Factor:0.999 - 1.001
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
About this item
:
:40 g/mol
Boiling Pt:105…140 °C (1.013 hPa)
Melting Pt:–12 – –10 °C.
Density:1.08 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1824
ADR:8,II
:GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548