Order Entry
Puerto Rico
ContactUsLinkComponent
Potassium permanganate 0.1 M KMnO4 (0.5N) volumetric solution, Fluka™
Irritant
Potassium permanganate 0.1 M KMnO4 (0.5N) volumetric solution, Fluka™
Catalog # BJ35201-2.5L
CAS Number:  
Irritant
Potassium permanganate 0.1 M KMnO4 (0.5N) volumetric solution, Fluka™
Catalog # BJ35201-2.5L
Supplier Number:  35201-4X2.5L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    2.5 L
  • Cat. No.:
    BJ35201-2.5L
  • Supplier no.:
    35201-2.5L

Specifications

Specification Test Results

  • Factor:
    0.999 - 1.001
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

 :
 :158.03 g/mol
Boiling Pt:100 °C (1.013 hPa)
Storage Temperature:Ambient
UN:3082
ADR:9,III
 :GHS07
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364