Order Entry
Puerto Rico
ContactUsLinkComponent
Potassium permanganate 0.2 M KMnO4 (1N) volumetric solution, Fluka™
Corrosive
Potassium permanganate 0.2 M KMnO4 (1N) volumetric solution, Fluka™
Catalog # BJ35184-1L
CAS Number:  
Corrosive
Potassium permanganate 0.2 M KMnO4 (1N) volumetric solution, Fluka™
Catalog # BJ35184-1L
Supplier Number:  35184-6X1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    BJ35184-1L
  • Supplier no.:
    35184-1L

Specifications

Specification Test Results

  • Factor:
    0.999 - 1.001
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

 :
 :158.03 g/mol
Storage Temperature:Ambient
UN:3082
ADR:9,III
 :GHS05
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364