
Zinc sulfate 0.05 M ZnSO4 ≥99% volumetric solution, Fluka™
BJ34448-1L
:
- Pack type:Plastic bottle
- Size:1 L
- Cat. No.:BJ34448-1L
- Supplier no.:34448-1L
- Factor:0.998 - 1.002
:
:100 °C (solvent)
:<0 °C
:1.005 g/cm³
: Does not flash
:287.56 g/mol
:GHS07
:Ambient
:7446-20-0
:ZnSO₄·7H₂O
:MFCD00149894