VeriSpec® Internal Calibration Standard, 7 Components, RICCA Chemical Company
Catalog # RCRV010862100N
Supplier: Ricca Chemical
CAS Number:
Specifications
- Pack type:Plastic Bottle
- Size:100 ml
- Cat. No.:RCRV010862100N
- Supplier no.:RV010862-100N
Specifications
About this item
Certified reference material (CRM) for ICP-MS
10 ppm: Bi, Ho, In, Li⁶⁺, Sc, Tb, Y
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility