Order Entry
Puerto Rico
ContactUsLinkComponent
VeriSpec™ Alternate Metals II, 4 Components, RICCA Chemical Company
VeriSpec™ Alternate Metals II, 4 Components, RICCA Chemical Company
Catalog # RCRV010856100N
Supplier:  Ricca Chemical
CAS Number:  
undefined
VeriSpec™ Alternate Metals II, 4 Components, RICCA Chemical Company
Catalog # RCRV010856100N
Supplier:  Ricca Chemical
Supplier Number:  RV010856-100N
CAS Number:  

Specifications

  • Pack type:
    Plastic Bottle
  • Size:
    100 ml
  • Cat. No.:
    RCRV010856100N
  • Supplier no.:
    RV010856-100N

Specifications

About this item

Certified reference material (CRM) for ICP-MS

500 ppm: Ca, Na; 100 ppm K, Mg

Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility