VeriSpec® ICP Tuning Solution, 15 components, RICCA Chemical Company
Supplier: Ricca Chemical
Certificates
About this item
Certified reference material (CRM) for ICP-MS
500 ppm: K; 50 ppm: Al, As, Ba, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Se, Sr, Zn
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility